Assay Details
Target Gene Details
Entrez Gene ID: | 16993 |
Gene Name: | leukotriene A4 hydrolase |
Gene Aliases: |
- |
Location: |
Chr.10:93453396-93484898 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lta4h | NM_008517.2 | NP_032543.2 | ||
| AK134931.1 | BAE22342.1 | |||
| AK180384.1 | ||||
| BC021417.1 | AAH21417.1 |

Genomic Map