Assay Details
Target Gene Details
Entrez Gene ID: | 18813 |
Gene Name: | proliferation-associated 2G4 |
Gene Aliases: |
38kDa, AA672939, Ebp1, Plfap |
Location: |
Chr.10:128557766-128565934 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pa2g4 | NM_011119.3 | NP_035249.1 | ||
| AK002685.1 | ||||
| AK076286.1 | ||||
| AK144645.1 | BAE25986.1 | |||
| AK166936.1 | BAE39129.1 | |||
| AK168582.1 | BAE40451.1 | |||
| AK205907.1 | ||||
| AK207779.1 | ||||
| BC003777.1 | ||||
| BC005593.1 | ||||
| BC028926.1 | ||||
| BC037623.1 | ||||
| BC046532.1 | AAH46532.1 | |||
| U43918.1 | AAB60513.1 | |||
| X84789.1 | CAA59260.1 |
More Information
Set Membership: |
|

Genomic Map