Assay Details
Target Gene Details
Entrez Gene ID: | 103199 |
Gene Name: | FIG4 homolog (S. cerevisiae) |
Gene Aliases: |
A530089I17Rik, AI326867, Sac3 |
Location: |
Chr.10:41188172-41303241 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fig4 | NM_133999.1 | 9 | 1192 | NP_598760.1 |
| AK041196.1 | 9 | 1193 | ||
| AK169881.1 | 9 | 1156 | BAE41433.1 | |
| AK172130.1 | 9 | 1186 | BAE42839.1 | |
| AK172341.1 | 9 | 1157 | ||
| AK172926.1 | 2 | 240 | BAD32204.1 | |
| BC015295.1 | 9 | 1192 | AAH15295.1 | |
| BC031887.1 | 9 | 1191 | AAH31887.1 |
More Information
Set Membership: |
|

Genomic Map