Assay Details
Target Gene Details
Entrez Gene ID: | 15182 |
Gene Name: | histone deacetylase 2 |
Gene Aliases: |
D10Wsu179e, YAF1, Yy1bp, mRPD3 |
Location: |
Chr.10:36974544-37001889 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac2 | NM_008229.2 | NP_032255.2 | ||
AB221663.1 | ||||
AK051753.1 | BAC34755.1 | |||
AK135572.1 | BAE22585.1 | |||
BC138517.1 | ||||
U31758.1 | AAC52889.1 |
