Assay Details
Target Gene Details
Entrez Gene ID: | 17937 |
Gene Name: | Ngfi-A binding protein 2 |
Gene Aliases: |
AI451907 |
Location: |
Chr.10:127660918-127666703 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nab2 | NM_001122895.1 | NP_001116367.1 | ||
| NM_008668.2 | NP_032694.2 | |||
| AK155501.1 | BAE33295.1 | |||
| AK158688.1 | BAE34612.1 | |||
| AK171180.1 | BAE42297.1 | |||
| BC045139.1 | AAH45139.1 | |||
| U47543.1 | AAC52650.1 |

Genomic Map