Assay Details
Target Gene Details
Entrez Gene ID: | 103466 |
Gene Name: | 5'-nucleotidase domain containing 3 |
Gene Aliases: |
AU040402, AW540062, C630002B14Rik, TU12B1-TY |
Location: |
Chr.10:86778990-86838389 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nt5dc3 | NM_175331.3 | 2 | 353 | NP_780540.2 |
| AK147488.1 | 2 | 322 | BAE27946.1 | |
| AK172248.1 | 2 | 310 | BAE42905.1 | |
| CJ098720.1 | 2 | 354 |
More Information
Set Membership: |
|

Genomic Map