Assay Details
Target Gene Details
Entrez Gene ID: | 17123 |
Gene Name: | mucosal vascular addressin cell adhesion molecule 1 |
Gene Aliases: |
AV211525 |
Location: |
Chr.10:79664395-79668541 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Madcam1 | NM_013591.2 | NP_038619.2 | ||
AK137028.1 | BAE23208.1 | |||
AK156096.1 | BAE33586.1 | |||
BC024372.1 | AAH24372.1 | |||
BC115910.1 | ||||
BC118056.1 | ||||
L21203.1 | AAA39485.1 | |||
S80258.1 | AAB35609.1 |
