Assay Details
Target Gene Details
Entrez Gene ID: | 100503659 |
Gene Name: | downstream of Stk11 |
Gene Aliases: |
9630008F14, R29144/1 |
Location: |
Chr.10:80130434-80140136 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dos | NM_001195268.1 | NP_001182197.1 | ||
| NR_036582.1 | ||||
| AK137009.1 | ||||
| BM947270.1 | ||||
| BY274870.1 | ||||
| BY280734.1 |
More Information
Set Membership: |
|

Genomic Map