Assay Details
Target Gene Details
Entrez Gene ID: | 14051 |
Gene Name: | eyes absent 4 homolog (Drosophila) |
Gene Aliases: |
B130023L16Rik |
Location: |
Chr.10:23102962-23350856 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eya4 | NM_010167.4 | 10 | 1026 | NP_034297.2 |
AK041625.1 | 10 | 1027 | BAC31011.1 | |
BC120898.1 | 10 | 1010 | ||
BC120899.1 | 10 | 1010 | ||
Y17115.1 | 11 | 1137 | CAA76637.1 |
