Assay Details
Target Gene Details
Entrez Gene ID: | 77976 |
Gene Name: | NUAK family, SNF1-like kinase, 1 |
Gene Aliases: |
AU014801, AW494241, B230104P22Rik, Omphk1 |
Location: |
Chr.10:84370900-84440471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nuak1 | NM_001004363.1 | NP_001004363.1 | ||
| AB182364.1 | BAD23995.1 | |||
| BC040467.1 | AAH40467.1 | |||
| BC082328.1 | AAH82328.1 |
More Information
Set Membership: |
|

Genomic Map