Assay Details
Target Gene Details
Entrez Gene ID: | 71207 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 4 |
Gene Aliases: |
4933436C10Rik, DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P |
Location: |
Chr.10:95547007-95564167 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt4 | NM_027722.4 | NP_081998.3 | ||
| AK017075.1 | BAB30582.1 | |||
| AK048062.1 | BAC33229.1 | |||
| AK182561.1 | ||||
| AK215744.1 | ||||
| AK216297.1 | ||||
| AK216517.1 | ||||
| BC027209.1 | AAH27209.1 | |||
| CT010342.1 | CAJ18550.1 |
More Information
Set Membership: |
|

Genomic Map