Assay Details
Target Gene Details
Entrez Gene ID: | 212898 |
Gene Name: | dermatan sulfate epimerase |
Gene Aliases: |
6030499O08, AI480506, B130024B19Rik, DS-epi1, Sart2 |
Location: |
Chr.10:34151386-34208275 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dse | NM_172508.2 | 5 | 1579 | NP_766096.1 |
AK031737.1 | 5 | 1266 | ||
AK045065.1 | 5 | 1486 | BAC32205.1 | |
AK150737.1 | 5 | 1743 | ||
AK153326.1 | 5 | 1252 | BAE31905.1 | |
BC053095.1 | 5 | 1579 | AAH53095.1 |
