Assay Details
Target Gene Details
Entrez Gene ID: | 52665 |
Gene Name: | enoyl Coenzyme A hydratase domain containing 1 |
Gene Aliases: |
1700028A24Rik, AI314462, AI930038, D10Ertd667e, MMCD |
Location: |
Chr.10:29311507-29346661 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Echdc1 | NM_001110195.1 | NP_001103665.1 | ||
| NM_025855.4 | NP_080131.4 | |||
| AK006444.1 | BAB24592.2 | |||
| AK016260.1 | ||||
| AK028775.1 | BAC26112.1 | |||
| AK150932.1 | BAE29969.1 | |||
| AK152285.1 | BAE31098.1 | |||
| AK153454.1 | BAE32007.1 | |||
| AK166589.1 | BAE38876.1 | |||
| AK166634.1 | BAE38907.1 | |||
| AK166660.1 | BAE38924.1 | |||
| BC066183.1 | AAH66183.1 | |||
| CJ051963.1 |
More Information
Set Membership: |
|

Genomic Map