Assay Details
Target Gene Details
Entrez Gene ID: | 14360 |
Gene Name: | Fyn proto-oncogene |
Gene Aliases: |
AI448320, AW552119 |
Location: |
Chr.10:39369770-39565381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fyn | NM_001122892.1 | NP_001116364.1 | ||
NM_001122893.1 | NP_001116365.1 | |||
NM_008054.2 | NP_032080.2 | |||
AK156584.1 | BAE33766.1 | |||
AK171646.1 | BAE42585.1 | |||
AK179168.1 | ||||
AK181277.1 | ||||
AK185949.1 | ||||
AK211878.1 | ||||
AK216658.1 | ||||
BC032149.1 | AAH32149.1 | |||
BC092217.1 | AAH92217.1 | |||
BQ179147.1 | ||||
M27266.1 | AAA37644.1 | |||
U70324.1 | AAB09568.1 |
