Assay Details
Target Gene Details
Entrez Gene ID: | 14360 |
Gene Name: | Fyn proto-oncogene |
Gene Aliases: |
AI448320, AW552119 |
Location: |
Chr.10:39369770-39565381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fyn | NM_001122892.1 | NP_001116364.1 | ||
| NM_001122893.1 | NP_001116365.1 | |||
| NM_008054.2 | NP_032080.2 | |||
| AK156584.1 | BAE33766.1 | |||
| AK171646.1 | BAE42585.1 | |||
| AK188892.1 | ||||
| BC032149.1 | AAH32149.1 | |||
| BC092217.1 | AAH92217.1 | |||
| BQ179147.1 | ||||
| M27266.1 | AAA37644.1 | |||
| U70324.1 | AAB09568.1 |

Genomic Map