Assay Details
Target Gene Details
Entrez Gene ID: | 327814 |
Gene Name: | protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 2 |
Gene Aliases: |
5330438O12, B230207K17Rik, E130120L08Rik, mKIAA4112 |
Location: |
Chr.10:106468872-106933468 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppfia2 | NM_001205341.1 | 20 | 2714 | NP_001192270.1 |
| NM_177373.4 | NP_796347.2 | |||
| AK220491.1 | 20 | 2671 | BAD90289.1 | |
| BC065133.1 | AAH65133.1 | |||
| EU568870.1 | 20 | 2685 |

Genomic Map