Assay Details
Target Gene Details
Entrez Gene ID: | 18537 |
Gene Name: | protein-L-isoaspartate (D-aspartate) O-methyltransferase 1 |
Gene Aliases: |
C79501, PIMT |
Location: |
Chr.10:7629380-7680973 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcmt1 | NM_008786.2 | NP_032812.2 | ||
| AK006162.1 | BAB24438.1 | |||
| AK075632.1 | BAC35869.1 | |||
| AK148789.1 | ||||
| AK208253.1 | ||||
| AK214928.1 | ||||
| BC049613.1 | AAH49613.1 | |||
| BC058966.1 | AAH58966.1 | |||
| D38023.1 | ||||
| M60320.1 | AAA92742.1 |
More Information
Set Membership: |
|

Genomic Map