Assay Details
Target Gene Details
Entrez Gene ID: | 268288 |
Gene Name: | sterile alpha motif domain containing 3 |
Gene Aliases: |
Gm623 |
Location: |
Chr.10:26229618-26272916 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Samd3 | NM_001013766.2 | 3 | 294 | NP_001013788.2 |
NM_001115154.1 | 3 | 294 | NP_001108626.1 | |
AK082201.1 | 3 | 295 | BAC38435.1 | |
AK144462.1 | 1 | 317 | ||
BC150876.1 | 3 | 295 |
