Assay Details
Target Gene Details
Entrez Gene ID: | 73075 |
Gene Name: | peptidylprolyl isomerase (cyclophilin)-like 6 |
Gene Aliases: |
2900084F20Rik |
Location: |
Chr.10:41490439-41514295 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppil6 | NM_028430.1 | NP_082706.1 | ||
| AI463820.1 | ||||
| AK013818.1 | BAB29003.1 | |||
| BC147756.1 | ||||
| BC147757.1 | ||||
| BP772786.1 |
More Information
Set Membership: |
|

Genomic Map