Assay Details
Target Gene Details
Entrez Gene ID: | 20598 |
Gene Name: | sphingomyelin phosphodiesterase 2, neutral |
Gene Aliases: |
AW108287, nSMase, nSMase1 |
Location: |
Chr.10:41487172-41490340 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smpd2 | NM_009213.2 | NP_033239.1 | ||
| AJ222800.1 | CAA10994.1 | |||
| AK075616.1 | BAC35861.1 | |||
| AK088962.1 | 1 | 789 | ||
| BC010978.1 | AAH10978.1 |
More Information
Set Membership: |
|

Genomic Map