Assay Details
Target Gene Details
Entrez Gene ID: | 19263 |
Gene Name: | protein tyrosine phosphatase, receptor type, B |
Gene Aliases: |
3230402H02Rik, C130094E24, Ptpz, Rptpb, VE-PTP, Veptp |
Location: |
Chr.10:116275516-116389538 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprb | NM_029928.2 | NP_084204.2 | ||
| AF157628.1 | AAF80346.1 | |||
| AK147439.1 | BAE27912.1 | |||
| AK147668.1 | BAE28060.1 | |||
| AK162366.1 | BAE36875.1 | |||
| AY077755.1 | AAL75813.1 | |||
| BC047086.1 | AAH47086.1 | |||
| BC141006.1 | ||||
| BC145111.1 | ||||
| X58289.1 |
More Information
Set Membership: |
|

Genomic Map