Assay Details
Target Gene Details
Entrez Gene ID: | 20397 |
Gene Name: | sphingosine phosphate lyase 1 |
Gene Aliases: |
AI428538, D10Xrf456, S1PL, Spl |
Location: |
Chr.10:61098642-61147686 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgpl1 | NM_009163.3 | 1 | 70 | NP_033189.2 |
| AK036747.1 | BAC29562.1 | |||
| AK037789.1 | BAC29872.1 | |||
| AK043024.1 | BAC31437.1 | |||
| AK049342.1 | BAC33695.1 | |||
| AK150053.1 | BAE29269.1 | |||
| AK173137.1 | BAD32415.1 | |||
| BC026135.1 | AAH26135.1 | |||
| BY061956.1 | 1 | 56 | ||
| BY075752.1 | 1 | 71 |

Genomic Map