Assay Details
Target Gene Details
Entrez Gene ID: | 353169 |
Gene Name: | solute carrier family 2 (facilitated glucose transporter), member 12 |
Gene Aliases: |
GLUT-12, Glut12 |
Location: |
Chr.10:22612723-22703880 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc2a12 | NM_178934.4 | 4 | 1832 | NP_849265.2 |
| AJ549317.2 | 4 | 1832 | CAD70577.1 | |
| AK028970.1 | 4 | 1805 | BAC26220.1 | |
| AK031659.1 | 4 | 1750 | BAC27497.1 | |
| AK035972.1 | 4 | 1713 | BAC29262.1 | |
| AK143263.1 | 4 | 1821 | BAE25329.1 | |
| AY230881.1 | 4 | 1731 | AAP45844.1 | |
| BC096454.1 | 4 | 1763 | AAH96454.1 | |
| BC116314.1 | 4 | 1693 | ||
| BC116315.1 | 4 | 1696 | ||
| BC138316.1 | 4 | 1679 | ||
| BC138317.1 | 4 | 1679 |

Genomic Map