Assay Details
Target Gene Details
Entrez Gene ID: | 215085 |
Gene Name: | solute carrier family 35, member F1 |
Gene Aliases: |
Duf6, E130016L02 |
Location: |
Chr.10:52690501-53111622 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc35f1 | NM_178675.4 | 2 | 714 | NP_848790.2 |
AK038601.1 | 2 | 709 | BAC30061.1 | |
AK048957.1 | 2 | 674 | BAC33499.1 | |
AK053426.1 | 2 | 683 | BAC35383.1 | |
AK053545.1 | 2 | 653 | BAC35422.1 | |
AK083768.1 | 2 | 653 | BAC39016.1 | |
AK139331.1 | 2 | 684 | BAE23959.1 | |
AK163806.1 | 2 | 514 | ||
AK165021.1 | 2 | 668 | BAE38004.1 | |
AK166268.1 | 2 | 641 | ||
BC059075.1 | 2 | 610 | AAH59075.1 |
