Assay Details
Target Gene Details
Entrez Gene ID: | 215085 |
Gene Name: | solute carrier family 35, member F1 |
Gene Aliases: |
Duf6, E130016L02 |
Location: |
Chr.10:52690501-53111622 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc35f1 | NM_178675.4 | NP_848790.2 | ||
AK038601.1 | BAC30061.1 | |||
AK048957.1 | BAC33499.1 | |||
AK053426.1 | BAC35383.1 | |||
AK053545.1 | BAC35422.1 | |||
AK083768.1 | BAC39016.1 | |||
AK139331.1 | BAE23959.1 | |||
AK163806.1 | ||||
AK165021.1 | BAE38004.1 | |||
AK166268.1 | ||||
BC059075.1 | AAH59075.1 |
