Assay Details
Target Gene Details
Entrez Gene ID: | 57319 |
Gene Name: | sphingomyelin phosphodiesterase, acid-like 3A |
Gene Aliases: |
0610010C24Rik, AI529588, ASM3A, ASML3, ASML3A |
Location: |
Chr.10:57794544-57811830 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smpdl3a | NM_020561.2 | NP_065586.3 | ||
| AK002455.1 | ||||
| AK151683.1 | BAE30607.1 | |||
| AK152282.1 | BAE31096.1 | |||
| AK185060.1 | ||||
| AK194580.1 | ||||
| AK196251.1 | ||||
| AK205531.1 | ||||
| BC139288.1 | ||||
| BC139289.1 | ||||
| Y08135.1 | CAA69329.1 |
More Information
Set Membership: |
|

Genomic Map