Assay Details
Target Gene Details
Entrez Gene ID: | 66399 |
Gene Name: | Ts translation elongation factor, mitochondrial |
Gene Aliases: |
2310050B20Rik, 9430024O13Rik, EF-TS, EF-Tsmt |
Location: |
Chr.10:127022332-127030814 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsfm | NM_025537.3 | NP_079813.1 | ||
| AJ277891.1 | CAC81913.1 | |||
| AK009898.1 | ||||
| AK012240.1 | BAB28113.1 | |||
| AK018591.1 | ||||
| AK020437.1 | BAB32099.1 | |||
| AK169164.1 | BAE40943.1 | |||
| AK171656.1 | BAE42591.1 | |||
| AK172112.1 | BAE42833.1 | |||
| AK215409.1 | ||||
| BC034286.1 | AAH34286.1 | |||
| BC051508.1 | AAH51508.1 | |||
| BC057904.1 | AAH57904.1 |
More Information
Set Membership: |
|

Genomic Map