Assay Details
Target Gene Details
Entrez Gene ID: | 66420 |
Gene Name: | polymerase (RNA) II (DNA directed) polypeptide E |
Gene Aliases: |
2410021N14Rik, 25kDa, AW208866, RPB5, XAP4 |
Location: |
Chr.10:80035953-80039659 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Polr2e | NM_025554.2 | 2 | 166 | NP_079830.2 |
| AK010567.1 | 2 | 174 | ||
| AK015823.1 | 2 | 164 | BAE43245.1 | |
| AK088610.1 | 2 | 222 | ||
| AK132109.1 | 2 | 221 | BAE20985.1 | |
| AK144668.1 | 2 | 227 | BAE25998.1 | |
| AK145201.1 | 2 | 230 | BAE26292.1 | |
| AK155368.1 | 2 | 199 | BAE33223.1 | |
| BC026842.1 | 2 | 157 | AAH26842.1 | |
| BC037681.1 | 2 | 221 | AAH37681.1 | |
| BC045521.1 | 2 | 166 | AAH45521.1 |
More Information
Set Membership: |
|

Genomic Map