Assay Details
Target Gene Details
Entrez Gene ID: | 19279 |
Gene Name: | protein tyrosine phosphatase, receptor type, R |
Gene Aliases: |
Gmcp1, PTP-SL, PTPBR7, RPTPRR, mPTP213 |
Location: |
Chr.10:116018200-116274932 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprr | NM_001161837.1 | 3 | 332 | NP_001155309.1 |
| NM_011217.2 | 4 | 938 | NP_035347.1 | |
| AK028054.1 | 4 | 939 | BAC25724.1 | |
| BC119230.1 | 4 | 871 | ||
| BC120547.1 | 4 | 871 | ||
| CJ061226.1 | 3 | 333 | ||
| D31898.1 | 4 | 901 | BAA06696.1 | |
| Z30313.1 | 3 | 327 | CAA82957.1 |

Genomic Map