Assay Details
Target Gene Details
Entrez Gene ID: | 19279 |
Gene Name: | protein tyrosine phosphatase, receptor type, R |
Gene Aliases: |
Gmcp1, PTP-SL, PTPBR7, RPTPRR, mPTP213 |
Location: |
Chr.10:116018200-116274932 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprr | NM_001161837.1 | 4 | 456 | NP_001155309.1 |
| NM_001161838.1 | 1 | 413 | NP_001155310.1 | |
| NM_001161839.1 | 2 | 104 | NP_001155311.1 | |
| NM_001161840.1 | 2 | 296 | NP_001155312.1 | |
| NM_011217.2 | 5 | 1062 | NP_035347.1 | |
| AF041866.1 | 2 | 221 | AAD09171.1 | |
| AF129509.1 | 1 | 337 | AAD29673.1 | |
| AK028054.1 | 5 | 1063 | BAC25724.1 | |
| BC119230.1 | 5 | 995 | ||
| BC120547.1 | 5 | 995 | ||
| CF914721.1 | 2 | 104 | ||
| CJ059486.1 | 1 | 353 | ||
| CJ061226.1 | ||||
| D31898.1 | 5 | 1025 | BAA06696.1 | |
| DV657487.1 | 2 | 296 | ||
| Z30313.1 | 4 | 451 | CAA82957.1 |

Genomic Map