Assay Details
Target Gene Details
Entrez Gene ID: | 108705 |
Gene Name: | pituitary tumor-transforming 1 interacting protein |
Gene Aliases: |
1810010L20Rik, AI314311, AU018448, C79540 |
Location: |
Chr.10:77581767-77598732 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pttg1ip | NM_145925.2 | NP_666037.1 | ||
AK035696.1 | BAC29156.1 | |||
AK075773.1 | BAC35947.1 | |||
AK088813.1 | BAC40589.1 | |||
AK089887.1 | BAC40985.1 | |||
AK098093.1 | BAC05231.1 | |||
AK151639.1 | BAE30569.1 | |||
AK152366.1 | BAE31156.1 | |||
AK159987.1 | BAE35537.1 | |||
AK167972.1 | BAE39966.1 | |||
AK168164.1 | BAE40125.1 | |||
AK170802.1 | BAE42037.1 | |||
AK170971.1 | BAE42150.1 | |||
AK170972.1 | BAE42151.1 | |||
AK171140.1 | BAE42273.1 | |||
BC023961.1 | 1 | 47 | AAH23961.1 | |
BC025533.1 | AAH25533.1 | |||
BC029144.1 | AAH29144.1 | |||
BC032184.1 | AAH32184.1 |
