Assay Details
Target Gene Details
Entrez Gene ID: | 19886 |
Gene Name: | Ros1 proto-oncogene |
Gene Aliases: |
Ros-1, c-ros |
Location: |
Chr.10:52044865-52195397 on Build GRCm38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ros1 | NM_011282.2 | 33 | 5573 | NP_035412.2 |
| U15443.1 | 33 | 5572 | AAA50600.1 | |
| X81650.1 | 33 | 5396 | CAA57310.1 |
More Information
Set Membership: |
|

Genomic Map