Assay Details
Target Gene Details
Entrez Gene ID: | 20431 |
Gene Name: | premelanosome protein |
Gene Aliases: |
D10H12S53E, D12S53Eh, Pmel17, Si, Silv, gp100, gp87 |
Location: |
Chr.10:128705168-128720238 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmel | NM_021882.4 | 4 | 427 | NP_068682.2 |
| AK012808.1 | 4 | 428 | BAB28486.1 | |
| AK131956.1 | 4 | 429 | BAE20900.1 | |
| AK184547.1 | 2 | 110 | ||
| AK204124.1 | 2 | 110 | ||
| AK217723.1 | 2 | 102 | ||
| BC082555.1 | 4 | 413 | AAH82555.1 | |
| U14133.1 | 4 | 402 | AAA69538.1 |
More Information
Set Membership: |
|

Genomic Map