Assay Details
Target Gene Details
Entrez Gene ID: | 20431 |
Gene Name: | premelanosome protein |
Gene Aliases: |
D10H12S53E, D12S53Eh, Pmel17, Si, Silv, gp100, gp87 |
Location: |
Chr.10:128705168-128720238 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmel | NM_021882.4 | NP_068682.2 | ||
| AF119092.1 | AAF19761.1 | |||
| AK012808.1 | BAB28486.1 | |||
| AK131956.1 | BAE20900.1 | |||
| AK178741.1 | ||||
| AK197730.1 | ||||
| BC082555.1 | AAH82555.1 | |||
| U14133.1 | AAA69538.1 |
More Information
Set Membership: |
|

Genomic Map