Assay Details
Target Gene Details
Entrez Gene ID: | 215789 |
Gene Name: | phosphatase and actin regulator 2 |
Gene Aliases: |
AV158170, BC012871 |
Location: |
Chr.10:13207717-13474521 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phactr2 | NM_001033257.4 | NP_001028429.1 | ||
| NM_001195065.1 | NP_001181994.1 | |||
| NM_001195066.1 | NP_001181995.1 | |||
| NM_001195096.1 | NP_001182025.1 | |||
| AK142930.1 | BAE25225.1 | |||
| BC012871.1 | AAH12871.1 | |||
| BC130264.1 |
More Information
Set Membership: |
|

Genomic Map