Assay Details
Target Gene Details
Entrez Gene ID: | 108705 |
Gene Name: | pituitary tumor-transforming 1 interacting protein |
Gene Aliases: |
1810010L20Rik, AI314311, AU018448, C79540 |
Location: |
Chr.10:77581767-77598732 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pttg1ip | NM_145925.2 | 4 | 367 | NP_666037.1 |
| AK035696.1 | 4 | 382 | BAC29156.1 | |
| AK075773.1 | 4 | 340 | BAC35947.1 | |
| AK088813.1 | 4 | 404 | BAC40589.1 | |
| AK089887.1 | 4 | 349 | BAC40985.1 | |
| AK098093.1 | 4 | 381 | BAC05231.1 | |
| AK151639.1 | 4 | 349 | BAE30569.1 | |
| AK152366.1 | 4 | 349 | BAE31156.1 | |
| AK159987.1 | 4 | 349 | BAE35537.1 | |
| AK167972.1 | 4 | 379 | BAE39966.1 | |
| AK168164.1 | 4 | 379 | BAE40125.1 | |
| AK170802.1 | 4 | 349 | BAE42037.1 | |
| AK170971.1 | 4 | 382 | BAE42150.1 | |
| AK170972.1 | 4 | 349 | BAE42151.1 | |
| AK171140.1 | 4 | 348 | BAE42273.1 | |
| AK191979.1 | 2 | 67 | ||
| BC023961.1 | 4 | 422 | AAH23961.1 | |
| BC025533.1 | 4 | 391 | AAH25533.1 | |
| BC029144.1 | 4 | 363 | AAH29144.1 | |
| BC032184.1 | 4 | 386 | AAH32184.1 |

Genomic Map