Assay Details
Target Gene Details
Entrez Gene ID: | 108705 |
Gene Name: | pituitary tumor-transforming 1 interacting protein |
Gene Aliases: |
1810010L20Rik, AI314311, AU018448, C79540 |
Location: |
Chr.10:77581767-77598732 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pttg1ip | NM_145925.2 | NP_666037.1 | ||
| AK035696.1 | BAC29156.1 | |||
| AK075773.1 | BAC35947.1 | |||
| AK088813.1 | BAC40589.1 | |||
| AK089887.1 | BAC40985.1 | |||
| AK098093.1 | BAC05231.1 | |||
| AK151639.1 | BAE30569.1 | |||
| AK152366.1 | BAE31156.1 | |||
| AK159987.1 | BAE35537.1 | |||
| AK167972.1 | BAE39966.1 | |||
| AK168164.1 | BAE40125.1 | |||
| AK170802.1 | BAE42037.1 | |||
| AK170971.1 | BAE42150.1 | |||
| AK170972.1 | BAE42151.1 | |||
| AK171140.1 | BAE42273.1 | |||
| AK178924.1 | ||||
| AK181674.1 | ||||
| AK193277.1 | ||||
| AK193680.1 | ||||
| AK196421.1 | ||||
| AK196698.1 | ||||
| AK199195.1 | ||||
| AK205669.1 | ||||
| AK216713.1 | ||||
| BC023961.1 | AAH23961.1 | |||
| BC025533.1 | AAH25533.1 | |||
| BC029144.1 | AAH29144.1 | |||
| BC032184.1 | AAH32184.1 |

Genomic Map