Assay Details
Target Gene Details
Entrez Gene ID: | 75089 |
Gene Name: | UHRF1 (ICBP90) binding protein 1-like |
Gene Aliases: |
2010319N22Rik, 4930506D01Rik, AA589495, C80672, E030041M21Rik, mKIAA0701 |
Location: |
Chr.10:89744991-89819891 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Uhrf1bp1l | NM_029166.2 | 11 | 1597 | NP_083442.2 |
AB093261.1 | 11 | 1562 | BAC41445.1 | |
AK087280.1 | 9 | 1149 | BAC39836.1 | |
AK141050.1 | 11 | 1606 | ||
BC132134.1 | 11 | 1390 | ||
BC138092.1 | 11 | 1390 |
