Assay Details
Target Gene Details
Entrez Gene ID: | 76719 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 |
Gene Aliases: |
1700081L11Rik, 9430041J06Rik, mKIAA1267 |
Location: |
Chr.11:104333229-104469507 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kansl1 | NM_001081045.1 | NP_001074514.1 | ||
| AK006970.1 | BAB24813.1 | |||
| AK122484.1 | BAC65766.1 | |||
| AK161032.1 | 2 | 262 | ||
| BC025052.1 | AAH25052.1 | |||
| BC043121.1 | ||||
| BC054752.1 | ||||
| BC079594.1 | AAH79594.1 |
More Information
Set Membership: |
|

Genomic Map