Assay Details
Target Gene Details
Entrez Gene ID: | 56321 |
Gene Name: | apoptosis antagonizing transcription factor |
Gene Aliases: |
4933415H02Rik, 5830465M17Rik, Che-1, Trb |
Location: |
Chr.11:84422856-84513570 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aatf | NM_019816.1 | NP_062790.1 | ||
| AF222801.1 | AAF26745.1 | |||
| AK016820.1 | ||||
| AK077789.1 | 9 | 1440 | BAC37011.1 | |
| AK141334.1 | BAE24653.1 | |||
| BC025080.1 | AAH25080.1 |
More Information
Set Membership: |
|

Genomic Map