Assay Details
Target Gene Details
Entrez Gene ID: | 11670 |
Gene Name: | aldehyde dehydrogenase family 3, subfamily A1 |
Gene Aliases: |
Ahd-4, Ahd4, Aldh, Aldh3 |
Location: |
Chr.11:61207537-61218418 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aldh3a1 | NM_001112725.1 | NP_001106196.1 | ||
| NM_007436.2 | NP_031462.2 | |||
| AF072815.1 | AAD15964.1 | |||
| AK144241.1 | BAE25792.1 | |||
| BC144946.1 | ||||
| BC150677.1 | ||||
| EL959277.1 | ||||
| U12785.1 | AAA20670.1 |
More Information
Set Membership: |
|

Genomic Map