Assay Details
Target Gene Details
Entrez Gene ID: | 14479 |
Gene Name: | ubiquitin specific peptidase 15 |
Gene Aliases: |
4921514G19Rik, AI327321, E430033I05Rik, Gcap18 |
Location: |
Chr.10:123105006-123197019 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp15 | NM_001301628.1 | NP_001288557.1 | ||
| NM_027604.4 | NP_081880.2 | |||
| AF468037.1 | AAL77418.1 | |||
| AK014891.1 | ||||
| AK122301.1 | BAC65583.1 | |||
| AK133852.1 | BAE21887.1 | |||
| AK145749.1 | BAE26626.1 | |||
| AK161469.1 | BAE36413.1 | |||
| AK161957.1 | ||||
| AK163315.1 | ||||
| AK168755.1 | BAE40593.1 | |||
| BC050042.1 | AAH50042.1 |
More Information
Set Membership: |
|

Genomic Map