Assay Details
Target Gene Details
Entrez Gene ID: | 68067 |
Gene Name: | RIKEN cDNA 3010026O09 gene |
Gene Aliases: |
2900016G09Rik, AI851514, AW475975 |
Location: |
Chr.11:50174851-50200115 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3010026O09Rik | NM_026543.3 | NP_080819.2 | ||
| AK003636.1 | BAB22904.1 | |||
| AK013536.1 | ||||
| AK019392.1 | BAB31698.1 | |||
| BC028428.1 | AAH28428.1 |

Genomic Map