Assay Details
Target Gene Details
Entrez Gene ID: | 140723 |
Gene Name: | calcium channel, voltage-dependent, gamma subunit 5 |
Gene Aliases: |
- |
Location: |
Chr.11:107874604-107915055 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacng5 | NM_001199301.1 | NP_001186230.1 | ||
| NM_080644.3 | NP_542375.1 | |||
| AF361347.1 | AAL50042.1 | |||
| AF458900.1 | AAM00597.1 | |||
| AK032260.1 | BAC27783.1 | |||
| AK032548.1 | BAC27920.1 | |||
| AK034065.1 | BAC28568.1 | |||
| AK080762.1 | BAC38012.1 | |||
| BC120894.1 | ||||
| BC120895.1 |
More Information
Set Membership: |
|

Genomic Map