Assay Details
Target Gene Details
Entrez Gene ID: | 268420 |
Gene Name: | alkB, alkylation repair homolog 5 (E. coli) |
Gene Aliases: |
AW050020, AW215868, Abh5, E130207K11, Ofoxd |
Location: |
Chr.11:60537683-60558512 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alkbh5 | NM_172943.4 | NP_766531.2 | ||
| AK049130.1 | ||||
| AK053695.1 | BAC35478.1 | |||
| AK053700.1 | BAC35481.1 | |||
| AK162072.1 | BAE36711.1 | |||
| AK163294.1 | BAE37281.1 | |||
| AK169700.1 | ||||
| BC052076.1 | AAH52076.1 |
More Information
Set Membership: |
|

Genomic Map