Assay Details
Target Gene Details
Entrez Gene ID: | 83767 |
Gene Name: | WAS protein family, member 1 |
Gene Aliases: |
AI195380, AI838537, Scar, WAVE, WAVE-1 |
Location: |
Chr.10:40883534-40938569 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wasf1 | NM_031877.3 | 2 | 153 | NP_114083.1 |
| AF290877.1 | 1 | 49 | AAG02214.1 | |
| AF467773.1 | 1 | 49 | AAL75574.1 | |
| BC016896.1 | 2 | 111 | AAH16896.1 | |
| BC053423.1 | 2 | 114 | AAH53423.1 | |
| BY332208.1 | 2 | 153 |
More Information
Set Membership: |
|

Genomic Map