Assay Details
Target Gene Details
Entrez Gene ID: | 69186 |
Gene Name: | transmembrane protein 256 |
Gene Aliases: |
1810027O10Rik, 3110009M16Rik |
Location: |
Chr.11:69838525-69839558 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem256 | NM_026982.1 | NP_081258.1 | ||
| AK007623.1 | ||||
| AK014035.1 | 1 | 104 | ||
| BC117077.1 | ||||
| BC117079.1 |

Genomic Map