Assay Details
Target Gene Details
Entrez Gene ID: | 104112 |
Gene Name: | ATP citrate lyase |
Gene Aliases: |
A730098H14Rik, AW538652 |
Location: |
Chr.11:100476352-100528000 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acly | NM_001199296.1 | NP_001186225.1 | ||
| NM_134037.3 | NP_598798.1 | |||
| AF332051.1 | AAK56080.1 | |||
| AF332052.1 | AAK56081.1 | |||
| AK043434.1 | ||||
| AK132755.1 | BAE21336.1 | |||
| AK135528.1 | ||||
| AK136812.1 | BAE23135.1 | |||
| AK149660.1 | BAE29010.1 | |||
| AK169703.1 | BAE41315.1 | |||
| BC021502.1 | AAH21502.1 | |||
| BC056378.1 | AAH56378.1 |
More Information
Set Membership: |
|

Genomic Map