Assay Details
Target Gene Details
Entrez Gene ID: | 11684 |
Gene Name: | arachidonate 12-lipoxygenase |
Gene Aliases: |
9930022G08Rik, Alox12p, P-12LO |
Location: |
Chr.11:70241455-70255386 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alox12 | NM_007440.4 | 8 | 1086 | NP_031466.2 |
| AK036898.1 | 8 | 1084 | BAC29629.1 | |
| AK036960.1 | 8 | 1042 | ||
| AK079232.1 | 8 | 1087 | BAC37583.1 | |
| AK087724.1 | 8 | 1087 | BAC39981.1 | |
| AK172216.1 | 8 | 1084 | BAE42887.1 | |
| AK172488.1 | 8 | 1087 | BAE43031.1 | |
| BC152329.1 | 8 | 1091 | ||
| BC152403.1 | 8 | 1091 | ||
| S80446.1 | 8 | 1098 | AAB36013.1 | |
| U04334.1 | 8 | 1017 | AAA20659.1 |
More Information
Set Membership: |
|

Genomic Map