Assay Details
Target Gene Details
Entrez Gene ID: | 109934 |
Gene Name: | active BCR-related gene |
Gene Aliases: |
- |
Location: |
Chr.11:76416732-76577739 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abr | NM_198894.2 | 1 | 165 | NP_942597.1 |
| BY126026.1 | 1 | 166 | ||
| CF535448.1 | 1 | 153 |
More Information
Set Membership: |
|

Genomic Map